Learn More
Waters Corp Waters Online Column, 80Å, 25 μm, 2.1 × 20 mm
Supplier:  Waters Corp 186000706
Encompass_Preferred
The Oasis HLB on-line cartridge column is designed for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE). It features a universal polymeric sorbent and is compatible with various narrow-bore analytical columns.
  • Designed for rugged, reproducible, ultra-fast Solid Phase Extraction (SPE).
  • Features a universal polymeric sorbent for broad analyte extraction.
  • Compatible with narrow-bore analytical columns and traditional HPLC fittings.
  • Fits into a Sentry™ Holder for quick replacement.
  • Water-wettable, reversed-phase sorbent with high capacity and enhanced polar analyte retention.
  • Manufactured in a cGMP, ISO 9001 certified facility, chromatographically tested for reliability.
  • Offers a wide operating pH range of 0 to 14 pH.
Catalog No. 50-780-471