Learn More
Waters Corp Waters Online Column, HLB, 15 μm, 3 mm, 20 mm
Supplier:  Waters Corp 186002038
Encompass_Preferred
The Oasis HLB on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. It contains the universal polymeric Oasis HLB sorbent, which is optimized for the extraction of a wide range of analytes from various matrices. This on-line column is designed for use with appropriate narrow-bore analytical columns, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry columns. The column features traditional HPLC column fittings and hardware.
  • Rugged, reproducible, and ultra-fast on-line analysis.
  • Wide choice of configurations, particle sizes, and sorbent chemistries.
  • Available with six patented sorbents—HLB, PRiME HLB, MCX, MAX, WCX, and WAX.
  • High recovery and reproducible results for a wide range of compounds.
  • Cartridge format for use with Spark Holland Prospekt-2™/Symbiosis™ systems is also available.
Catalog No. 50-780-894