Learn More
Waters Corp Waters Oasis HLB Online Column, 80Å, 25 μm, 3 mm × 20 mm
Catalog No. 50780895 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB on-line column is engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE). It features the universal polymeric HLB sorbent, optimized for the extraction of a wide range of analytes from various matrices. This column is designed for use with appropriate narrow-bore analytical columns and includes traditional HPLC column fittings and hardware.
  • Features HLB chemistry for versatile reversed phase separation.
  • Constructed with a spherical copolymer particle.
  • Exceptionally stable across a pH range of 0 to 14.
  • Has a particle size of 25 μm, pore size of 80 Å, 3 mm inner diameter, and 20 mm length.
  • Water-wettable sorbent allows simplified 3-step SPE protocols, reducing solvent use by up to 70% and saving 40% sample prep time.
  • Effectively retains diverse compounds without conditioning or equilibration steps.
Catalog No. 50-780-895 Supplier Waters Corp Supplier No. 186002039
The Oasis HLB on-line column is engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE). It features the universal polymeric HLB sorbent, optimized for the extraction of a wide range of analytes from various matrices. This column is designed for use with appropriate narrow-bore analytical columns and includes traditional HPLC column fittings and hardware.
  • Features HLB chemistry for versatile reversed phase separation.
  • Constructed with a spherical copolymer particle.
  • Exceptionally stable across a pH range of 0 to 14.
  • Has a particle size of 25 μm, pore size of 80 Å, 3 mm inner diameter, and 20 mm length.
  • Water-wettable sorbent allows simplified 3-step SPE protocols, reducing solvent use by up to 70% and saving 40% sample prep time.
  • Effectively retains diverse compounds without conditioning or equilibration steps.