Learn More
Waters Corp Waters Oasis HLB Online Column, 80Å, 5 μm, 3.9 mm × 20 mm
Catalog No. 50780896 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB on-line column is engineered for rugged, reproducible, ultra-fast Solid Phase Extraction (SPE). It uses a universal polymeric sorbent, optimized for extracting diverse analytes. Compatible with narrow-bore analytical columns, it features standard HPLC fittings. This multipurpose reverse-phase sorbent enhances sensitivity and specificity via mixed-mode capabilities. Its water-wettable nature eliminates pre-conditioning, cutting solvent use and preparation time.
  • Rugged, reproducible, ultra-fast on-line SPE.
  • Universal polymeric sorbent extracts diverse analytes.
  • Compatible with narrow-bore analytical columns.
  • Standard HPLC fittings and hardware.
  • Multipurpose reverse-phase sorbent enhances sensitivity, specificity.
  • Water-wettable, for direct aqueous sample loading.
  • Eliminates equilibration/conditioning, reduces solvent/prep time.
Catalog No. 50-780-896 Supplier Waters Corp Supplier No. 186002040
The Oasis HLB on-line column is engineered for rugged, reproducible, ultra-fast Solid Phase Extraction (SPE). It uses a universal polymeric sorbent, optimized for extracting diverse analytes. Compatible with narrow-bore analytical columns, it features standard HPLC fittings. This multipurpose reverse-phase sorbent enhances sensitivity and specificity via mixed-mode capabilities. Its water-wettable nature eliminates pre-conditioning, cutting solvent use and preparation time.
  • Rugged, reproducible, ultra-fast on-line SPE.
  • Universal polymeric sorbent extracts diverse analytes.
  • Compatible with narrow-bore analytical columns.
  • Standard HPLC fittings and hardware.
  • Multipurpose reverse-phase sorbent enhances sensitivity, specificity.
  • Water-wettable, for direct aqueous sample loading.
  • Eliminates equilibration/conditioning, reduces solvent/prep time.