Learn More
Waters Corp Waters Oasis HLB Online Column, 80Å, 5 μm, 3 mm X 20 mm
Supplier:  Waters Corp 186002037
Encompass_Preferred
The Oasis HLB on-line column is designed for rugged, reproducible, and ultra-fast on-line solid-phase extraction (SPE). It contains the universal polymeric Oasis HLB sorbent, optimized for extracting a wide range of analytes from various matrices. This column is intended for use with appropriate Waters narrow-bore analytical columns and features traditional HPLC column fittings and hardware.
  • Accelerates solid-phase extraction for easy integration into LC systems.
  • Ensures high recovery and reproducible results for diverse compounds.
  • Isolates acidic, basic, and neutral compounds from various matrices.
  • Simplifies sample preparation and reduces guesswork with a generic protocol.
  • Utilizes universal polymeric HLB sorbent for broad analyte extraction.
  • Equipped with traditional HPLC column fittings and hardware.
Catalog No. 50-780-893