Learn More
Waters Corp Online Column, Oasis HLB, 5μm, 4.6 × 20 mm
Supplier:  Waters Corp 186002043
Encompass_Preferred
The Oasis HLB on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. It contains the universal polymeric HLB sorbent, optimized for the extraction of a wide range of analytes from various matrices. The column is intended for use with appropriate narrow-bore analytical columns, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry columns. It features traditional HPLC column fittings and hardware.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE.
  • Contains the universal polymeric HLB sorbent.
  • Optimized for the extraction of a wide range of analytes from various matrices.
  • Intended for use with appropriate narrow-bore analytical columns.
  • Features traditional HPLC column fittings and hardware.
Catalog No. 50-780-899