Learn More
Waters Corp Waters Oasis HLB 96-well Plate, 10 mg Sorbent, 30 μm
Catalog No. 50780079 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB 96-well plate utilizes a universal polymeric reversed-phase sorbent, ideal for extracting acidic, basic, and neutral compounds from various matrices. Its water-wettable nature ensures consistent results, even if wells dry out. It is compatible with many manifold configurations and robotic liquid handling systems.
  • Ensures consistent results even if wells dry out before sample loading.
  • Suitable for high throughput and high recovery applications.
  • Innovative two-stage well design aids clean extracts, low matrix effects, and method variability.
  • Manufactured in ISO 9000 registered facilities under cGMP guidelines for reproducibility.
  • Sorbent is stable from pH 0 to 14.
  • Simplifies protocols by reducing processing steps, solvent consumption, and sample preparation time.
  • Maintains high recoveries for diverse compounds without requiring conditioning and equilibration.
Catalog No. 50-780-079 Supplier Waters Corp Supplier No. 186000128
The Oasis HLB 96-well plate utilizes a universal polymeric reversed-phase sorbent, ideal for extracting acidic, basic, and neutral compounds from various matrices. Its water-wettable nature ensures consistent results, even if wells dry out. It is compatible with many manifold configurations and robotic liquid handling systems.
  • Ensures consistent results even if wells dry out before sample loading.
  • Suitable for high throughput and high recovery applications.
  • Innovative two-stage well design aids clean extracts, low matrix effects, and method variability.
  • Manufactured in ISO 9000 registered facilities under cGMP guidelines for reproducibility.
  • Sorbent is stable from pH 0 to 14.
  • Simplifies protocols by reducing processing steps, solvent consumption, and sample preparation time.
  • Maintains high recoveries for diverse compounds without requiring conditioning and equilibration.