Learn More
Waters Corp Waters Oasis HLB 96-well Plate, 60 mg Sorbent, 60 μm
Catalog No. 50818654 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB 96-well plate utilizes a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. Its unique balance of hydrophobicity and water-wettability prevents issues if wells dry out. With a 60 μm particle size, it is ideal for viscous samples and compatible with various manifold configurations and robotic liquid handling systems.
  • Innovative, two-stage well design offers high throughput and high recovery.
  • Easily automated for reliable, high-throughput solid phase extraction (SPE).
  • 60 mg capacity per well allows for storing more analytes without splitting samples.
  • Water-wettable quality ensures robust methods and outstanding retention capacity.
  • Fast, reliable generic method for LC/MS/MS analysis, yielding high recoveries (>85%) and consistent results (<5%RSD).
Catalog No. 50-818-654 Supplier Waters Corp Supplier No. 186000679
The Oasis HLB 96-well plate utilizes a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. Its unique balance of hydrophobicity and water-wettability prevents issues if wells dry out. With a 60 μm particle size, it is ideal for viscous samples and compatible with various manifold configurations and robotic liquid handling systems.
  • Innovative, two-stage well design offers high throughput and high recovery.
  • Easily automated for reliable, high-throughput solid phase extraction (SPE).
  • 60 mg capacity per well allows for storing more analytes without splitting samples.
  • Water-wettable quality ensures robust methods and outstanding retention capacity.
  • Fast, reliable generic method for LC/MS/MS analysis, yielding high recoveries (>85%) and consistent results (<5%RSD).