Learn More
Waters Corp Waters Corporation Oasis HLB Plus Short Cartridge, 225 mg Sorbent, 60 μm, Reversed Phase
Catalog No. 50780080 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis HLB Plus Short cartridges use Oasis HLB, a universal polymeric reversed-phase sorbent, for extracting diverse compounds from various matrices. Water wettable, ensures high retention and recovery even if dry. With 60 μm particle size, they suit viscous samples and integrate easily with pumps, syringes, or vacuum-assisted and automated systems.
  • Oasis HLB, universal polymeric reversed-phase sorbent.
  • Water wettable for consistent retention, even when dry.
  • Sorbent weight: 225 mg; Particle size: 60 μm.
  • Chemistry: HLB; Mode: Reversed Phase.
  • Pore size: 80 Å; Substrate: Copolymer.
  • pH range: 0–14; Material: Polypropylene.
  • Package includes 50 cartridges.
  • Extracts acidic, basic, and neutral compounds.
  • Compatible with positive-pressure, vacuum-assisted systems.
  • Improved recovery and reproducibility via enhanced retention.
Catalog No. 50-780-080 Supplier Waters Corp Supplier No. 186000132
Oasis HLB Plus Short cartridges use Oasis HLB, a universal polymeric reversed-phase sorbent, for extracting diverse compounds from various matrices. Water wettable, ensures high retention and recovery even if dry. With 60 μm particle size, they suit viscous samples and integrate easily with pumps, syringes, or vacuum-assisted and automated systems.
  • Oasis HLB, universal polymeric reversed-phase sorbent.
  • Water wettable for consistent retention, even when dry.
  • Sorbent weight: 225 mg; Particle size: 60 μm.
  • Chemistry: HLB; Mode: Reversed Phase.
  • Pore size: 80 Å; Substrate: Copolymer.
  • pH range: 0–14; Material: Polypropylene.
  • Package includes 50 cartridges.
  • Extracts acidic, basic, and neutral compounds.
  • Compatible with positive-pressure, vacuum-assisted systems.
  • Improved recovery and reproducibility via enhanced retention.