Learn More
Waters Corp Oasis Plus Light Cartridge, 30 mg Sorbent, 30 μm Particle Size, HLB
Catalog No. 50466126 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Plus Light Cartridge features a polymeric, water-wettable reversed-phase sorbent, designed for the extraction of a wide range of compounds from various matrices with high, reproducible recoveries. It functions as a Hydrophilic-Lipophilic-Balanced (HLB) universal sorbent for acidic, neutral, and basic materials.
  • Polymeric, water-wettable reversed-phase sorbent.
  • Hydrophilic-Lipophilic-Balanced (HLB) design.
  • Universal sorbent for acidic, neutral, and basic materials.
  • Exceptional retention for polar molecules.
  • Three times higher hydrophobic retention capacity than typical silica-based SPE sorbents.
  • Maintains good retention capacity even if the sorbent bed runs dry.
  • Works across a wide pH range of 0 to 14.
  • Designed for use with vacuum manifolds and automated SPE devices.
Catalog No. 50-466-126 Supplier Waters Corp Supplier No. 186005125
The Plus Light Cartridge features a polymeric, water-wettable reversed-phase sorbent, designed for the extraction of a wide range of compounds from various matrices with high, reproducible recoveries. It functions as a Hydrophilic-Lipophilic-Balanced (HLB) universal sorbent for acidic, neutral, and basic materials.
  • Polymeric, water-wettable reversed-phase sorbent.
  • Hydrophilic-Lipophilic-Balanced (HLB) design.
  • Universal sorbent for acidic, neutral, and basic materials.
  • Exceptional retention for polar molecules.
  • Three times higher hydrophobic retention capacity than typical silica-based SPE sorbents.
  • Maintains good retention capacity even if the sorbent bed runs dry.
  • Works across a wide pH range of 0 to 14.
  • Designed for use with vacuum manifolds and automated SPE devices.