Learn More
Waters Corp Waters Oasis HLB Vac RC Cartridge, 20 cc, 30 mg Sorbent, 30 μm
Catalog No. 50780203 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Vac RC cartridges use Oasis HLB sorbent, a water-wettable, polymeric reversed-phase sorbent for efficient extraction of diverse compounds. It ensures high retention and recoveries, even when dry. An integral 20 cc funnel-shaped reservoir accommodates larger samples and is compatible with robotic systems. The sorbent offers pH extreme stability, broad solvent compatibility, and enhanced hydrophobic retention for improved recovery and reproducibility.
  • Solid Phase Extraction product type
  • Vac RC Cartridge format
  • 30 mg sorbent weight
  • 30 μm sorbent particle size
  • 20 cc cartridge volume
  • 50 units per package
  • HLB chemistry
  • Not endcapped
  • Mass spectrometer compatible
  • Reversed phase mode
  • 80 Å pore size
  • Copolymer sorbent substrate
  • Water wettable
  • pH range 0 to 14
  • Polypropylene material
Catalog No. 50-780-203 Supplier Waters Corp Supplier No. 186000382
These Vac RC cartridges use Oasis HLB sorbent, a water-wettable, polymeric reversed-phase sorbent for efficient extraction of diverse compounds. It ensures high retention and recoveries, even when dry. An integral 20 cc funnel-shaped reservoir accommodates larger samples and is compatible with robotic systems. The sorbent offers pH extreme stability, broad solvent compatibility, and enhanced hydrophobic retention for improved recovery and reproducibility.
  • Solid Phase Extraction product type
  • Vac RC Cartridge format
  • 30 mg sorbent weight
  • 30 μm sorbent particle size
  • 20 cc cartridge volume
  • 50 units per package
  • HLB chemistry
  • Not endcapped
  • Mass spectrometer compatible
  • Reversed phase mode
  • 80 Å pore size
  • Copolymer sorbent substrate
  • Water wettable
  • pH range 0 to 14
  • Polypropylene material