Learn More
Waters Corp Waters Direct Connect Column, HLB, 20 μm, 2.1 mm x 30 mm
Catalog No. NC1669049 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB direct connect column is designed for rugged, reproducible, and ultra-fast on-line SPE. It contains the universal polymeric HLB sorbent, which is optimized for the extraction of a wide range of analytes from various matrices. It is intended to be used with appropriate narrow-bore analytical columns.
  • Direct connect hardware provides convenient column connection.
  • Hydrophilic-Lipophilic-Balanced (HLB), water-wettable, reversed-phase sorbent.
  • Composed of N-vinylpyrrolidone and divinylbenzene monomers.
  • Superior reversed-phase capacity with neutral polar ‘hook’ for polar analytes.
  • Water-wettable sorbents ensure excellent retention capacity for a wider polarity spectrum, even if dry.
  • Connects directly to a switching valve or like a conventional HPLC column.
  • High retention capacity improves recovery and reproducibility.
Catalog No. NC1669049 Supplier Waters Corp Supplier No. 186005231
The Oasis HLB direct connect column is designed for rugged, reproducible, and ultra-fast on-line SPE. It contains the universal polymeric HLB sorbent, which is optimized for the extraction of a wide range of analytes from various matrices. It is intended to be used with appropriate narrow-bore analytical columns.
  • Direct connect hardware provides convenient column connection.
  • Hydrophilic-Lipophilic-Balanced (HLB), water-wettable, reversed-phase sorbent.
  • Composed of N-vinylpyrrolidone and divinylbenzene monomers.
  • Superior reversed-phase capacity with neutral polar ‘hook’ for polar analytes.
  • Water-wettable sorbents ensure excellent retention capacity for a wider polarity spectrum, even if dry.
  • Connects directly to a switching valve or like a conventional HPLC column.
  • High retention capacity improves recovery and reproducibility.