Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Oasis MAX Cartridge, 6 cc, 400 mg, 30 μm, Flangeless Vac

Catalog No. 50179318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Oasis MAX 6 cc Flangeless Vac Cartridge is designed for solid phase extraction (SPE) applications. It features 400 mg of sorbent with a 30 μm particle size, housed in a 6 cc polypropylene barrel. This mixed-mode, water-wettable cartridge is compatible with mass spectrometry and offers a broad pH range from 0 to 14, providing reliable performance for various sample preparation needs.

  • Format as a flangeless vac cartridge.
  • Contains 400 mg of sorbent.
  • Features a particle size of 30 μm.
  • Designed with a pore size of 80 Å.
  • Utilizes a copolymer sorbent substrate.
  • Is water wettable, ensuring consistent performance.
  • Offers a broad pH range of 0 to 14.
  • Compatible with mass spectrometry.
  • Constructed from polypropylene material.
  • Has a barrel size of 6 cc.

Catalog No. 50-179-318 Supplier Waters Corp Supplier No. 186001855
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Oasis MAX 6 cc Flangeless Vac Cartridge is designed for solid phase extraction (SPE) applications. It features 400 mg of sorbent with a 30 μm particle size, housed in a 6 cc polypropylene barrel. This mixed-mode, water-wettable cartridge is compatible with mass spectrometry and offers a broad pH range from 0 to 14, providing reliable performance for various sample preparation needs.

  • Format as a flangeless vac cartridge.
  • Contains 400 mg of sorbent.
  • Features a particle size of 30 μm.
  • Designed with a pore size of 80 Å.
  • Utilizes a copolymer sorbent substrate.
  • Is water wettable, ensuring consistent performance.
  • Offers a broad pH range of 0 to 14.
  • Compatible with mass spectrometry.
  • Constructed from polypropylene material.
  • Has a barrel size of 6 cc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.