Learn More
Waters Corp Waters Oasis MAX μElution Plate, 96-well, 2 mg, 30 μm
Catalog No. 50818607 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This innovative 96-well μElution plate is designed for SPE cleanup and analyte enrichment, handling sample volumes from 10 μL to 375 μL. Each well contains 2 mg of a novel, mixed-mode polymeric sorbent, optimized for extracting acidic compounds with high selectivity and sensitivity. Elution is possible in only 25 μL, providing sensitive, robust, and reproducible results with a 15-fold sample enrichment, eliminating the need for evaporation and reconstitution. The plate is compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Strong, water-wettable, polymeric sorbent stable from pH 0 to 14, preventing wells from drying out.
  • Separates analytes into acidic and basic/neutral fractions.
  • Reproducible delivery with 0.25 meq/g quaternary-amine-ion-exchange capacity.
  • Innovative two-stage well design for high throughput and recovery.
Catalog No. 50-818-607 Supplier Waters Corp Supplier No. 186001829
This innovative 96-well μElution plate is designed for SPE cleanup and analyte enrichment, handling sample volumes from 10 μL to 375 μL. Each well contains 2 mg of a novel, mixed-mode polymeric sorbent, optimized for extracting acidic compounds with high selectivity and sensitivity. Elution is possible in only 25 μL, providing sensitive, robust, and reproducible results with a 15-fold sample enrichment, eliminating the need for evaporation and reconstitution. The plate is compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Strong, water-wettable, polymeric sorbent stable from pH 0 to 14, preventing wells from drying out.
  • Separates analytes into acidic and basic/neutral fractions.
  • Reproducible delivery with 0.25 meq/g quaternary-amine-ion-exchange capacity.
  • Innovative two-stage well design for high throughput and recovery.