Learn More
Waters Corp Waters Vac Cartridge, Oasis MAX, 1 cc, 30 mg, 30 μm
Catalog No. 50818756 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc cartridges feature a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds.
  • Novel, mixed-mode polymeric sorbent with anion-exchange groups.
  • Optimized for high selectivity and sensitivity for acidic compound extraction.
  • Water wettable, ensuring high retention and recoveries even when dry.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Simplifies SPE, providing clean samples and high recovery.
Catalog No. 50-818-756 Supplier Waters Corp Supplier No. 186000366
These 1 cc cartridges feature a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds.
  • Novel, mixed-mode polymeric sorbent with anion-exchange groups.
  • Optimized for high selectivity and sensitivity for acidic compound extraction.
  • Water wettable, ensuring high retention and recoveries even when dry.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Simplifies SPE, providing clean samples and high recovery.