Learn More
Waters Corp Oasis MAX Vac Cartridge, 3 cc, 60 mg, 60 μm
Catalog No. 50818660 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX 3 cc Vac Cartridge features a novel, mixed-mode polymeric sorbent, ideal for extracting acidic compounds with high selectivity and sensitivity. Its water-wettable sorbent ensures robust performance, even if dry, and its 60 μm particle size is suitable for viscous samples. This cartridge integrates seamlessly with vacuum manifolds and automated SPE instruments.
  • Optimized polymeric sorbent for high selectivity and sensitivity in acidic compound extraction.
  • Water-wettable sorbent ensures high retention and recoveries even when dry.
  • 60 μm particle size recommended for viscous samples.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Provides stability across a 0-14 pH range.
  • Removes phospholipids and lysophospholipids to reduce LC/MS/MS ion suppression.
Catalog No. 50-818-660 Supplier Waters Corp Supplier No. 186000368
The Oasis MAX 3 cc Vac Cartridge features a novel, mixed-mode polymeric sorbent, ideal for extracting acidic compounds with high selectivity and sensitivity. Its water-wettable sorbent ensures robust performance, even if dry, and its 60 μm particle size is suitable for viscous samples. This cartridge integrates seamlessly with vacuum manifolds and automated SPE instruments.
  • Optimized polymeric sorbent for high selectivity and sensitivity in acidic compound extraction.
  • Water-wettable sorbent ensures high retention and recoveries even when dry.
  • 60 μm particle size recommended for viscous samples.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Provides stability across a 0-14 pH range.
  • Removes phospholipids and lysophospholipids to reduce LC/MS/MS ion suppression.