Learn More
Waters Corp Oasis MAX 6 cc Vac Cartridge, 150 mg Sorbent per Cartridge, 60 µm, 30/pk
Catalog No. 50818757 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 6 cc Oasis MAX cartridges contain the Oasis MAX, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting acidic compounds with anion-exchange groups. Since the Oasis MAX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading. The 60 µm particle size is recommended when working with viscous samples. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-818-757 Supplier Waters Corp Supplier No. 186000370
The 6 cc Oasis MAX cartridges contain the Oasis MAX, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting acidic compounds with anion-exchange groups. Since the Oasis MAX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading. The 60 µm particle size is recommended when working with viscous samples. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.