Learn More
Waters Corp Oasis MAX Vac Cartridge, 6 cc, 500 mg, 60 μm
Catalog No. 50818661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 6 cc Vac cartridges utilize a novel, water-wettable, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds. Designed for viscous samples, they integrate seamlessly with vacuum manifolds and automated Solid Phase Extraction (SPE) instruments.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water-wettable sorbent maintains high retention and recovery even if dried.
  • 60 μm particle size is ideal for viscous samples.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Stable across a pH range of 0 to 14.
  • Reduces chromatographic complexity and enhances signal-to-noise ratio.
  • Improves detection limits and minimizes matrix effects.
Catalog No. 50-818-661 Supplier Waters Corp Supplier No. 186000865
These 6 cc Vac cartridges utilize a novel, water-wettable, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds. Designed for viscous samples, they integrate seamlessly with vacuum manifolds and automated Solid Phase Extraction (SPE) instruments.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water-wettable sorbent maintains high retention and recovery even if dried.
  • 60 μm particle size is ideal for viscous samples.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Stable across a pH range of 0 to 14.
  • Reduces chromatographic complexity and enhances signal-to-noise ratio.
  • Improves detection limits and minimizes matrix effects.