Learn More
Waters Corp Waters Vac Cartridge, Oasis MAX, 6 cc, 150 mg, 30 μm
Catalog No. 50780194 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MAX 6 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for higher selectivity and sensitivity in extracting acidic compounds. This water wettable sorbent maintains excellent recoveries even if it runs dry. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
  • Optimized for higher selectivity and sensitivity with acidic compounds.
  • Water wettable sorbent ensures high retention and excellent recoveries even when dry.
  • Ensures reproducible SPE protocols for acidic compounds and metabolites.
  • Stable from pH 0 to 14, facilitating fast method development.
  • Enables separation of analytes into two distinct fractions.
  • Compatible with vacuum manifolds and automated SPE instruments.
Catalog No. 50-780-194 Supplier Waters Corp Supplier No. 186000369
Oasis MAX 6 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for higher selectivity and sensitivity in extracting acidic compounds. This water wettable sorbent maintains excellent recoveries even if it runs dry. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
  • Optimized for higher selectivity and sensitivity with acidic compounds.
  • Water wettable sorbent ensures high retention and excellent recoveries even when dry.
  • Ensures reproducible SPE protocols for acidic compounds and metabolites.
  • Stable from pH 0 to 14, facilitating fast method development.
  • Enables separation of analytes into two distinct fractions.
  • Compatible with vacuum manifolds and automated SPE instruments.