Learn More
Waters Corp Waters SPE Cartridge, 1 cc, 30 mg, 30 μm, Flangeless
Catalog No. 50780787 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MAX solid phase extraction products are mixed-mode anion eXchange sorbents designed for acids. Built upon the unique water-wettable Oasis HLB Copolymer, these products offer both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity and sensitivity, providing clean samples with high sensitivity and reduced matrix effects for LC-MS/MS analyses.
  • Limits the impact of matrix interferences on sensitivity and increases the robustness and reproducibility of analytical results
  • Combines reversed-phase and ion exchange retention for greater cleanup selectivity and sensitivity for acidic compounds
  • Ensures high batch-to-batch and lot-to-lot reproducibility for robust, accurate, and precise analytical methods
  • Available in a 1 cc flangeless cartridge format with 30 mg sorbent and 30 μm particle size
Catalog No. 50-780-787 Supplier Waters Corp Supplier No. 186001883
Oasis MAX solid phase extraction products are mixed-mode anion eXchange sorbents designed for acids. Built upon the unique water-wettable Oasis HLB Copolymer, these products offer both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity and sensitivity, providing clean samples with high sensitivity and reduced matrix effects for LC-MS/MS analyses.
  • Limits the impact of matrix interferences on sensitivity and increases the robustness and reproducibility of analytical results
  • Combines reversed-phase and ion exchange retention for greater cleanup selectivity and sensitivity for acidic compounds
  • Ensures high batch-to-batch and lot-to-lot reproducibility for robust, accurate, and precise analytical methods
  • Available in a 1 cc flangeless cartridge format with 30 mg sorbent and 30 μm particle size