Learn More
Waters Corp Oasis MAX Flangeless Vac Cartridge 3 cc, 60 mg, 60 μm
Catalog No. 50780788 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MAX 3 cc Flangeless Vac Cartridges utilize a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds. The water-wettable sorbent ensures high retention and excellent recoveries even if dry, removing the need for special precautions. Its flangeless design allows for close packing in automated SPE systems and it supports a 0–14 pH range.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water-wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Flangeless configuration enables close packing for automated SPE systems.
  • Separates acidic and basic/neutral chemicals and effectively removes phospholipids and lysophospholipids.
  • 60 μm particle size is recommended for viscous samples.
  • Each pack contains 100 cartridges.
Catalog No. 50-780-788 Supplier Waters Corp Supplier No. 186001884
Oasis MAX 3 cc Flangeless Vac Cartridges utilize a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in extracting acidic compounds. The water-wettable sorbent ensures high retention and excellent recoveries even if dry, removing the need for special precautions. Its flangeless design allows for close packing in automated SPE systems and it supports a 0–14 pH range.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water-wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Flangeless configuration enables close packing for automated SPE systems.
  • Separates acidic and basic/neutral chemicals and effectively removes phospholipids and lysophospholipids.
  • 60 μm particle size is recommended for viscous samples.
  • Each pack contains 100 cartridges.