Learn More
Waters Corp Waters Oasis MAX Flangeless Vac Cartridge, 1 cc, 10 mg, 30 μm
Catalog No. 50201606 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc flangeless Oasis MAX cartridges contain a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need to take extraordinary steps to keep sorbent beds from drying out before sample loading.
  • Flangeless configuration allows the cartridges to be closely packed for processing in specialized automated SPE systems.
Catalog No. 50-201-606 Supplier Waters Corp Supplier No. 186006341
The 1 cc flangeless Oasis MAX cartridges contain a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need to take extraordinary steps to keep sorbent beds from drying out before sample loading.
  • Flangeless configuration allows the cartridges to be closely packed for processing in specialized automated SPE systems.