Learn More
Waters Corp Waters Corporation Cartridge, Oasis MAX Online, 80Å, 30 μm, 10 mm
Catalog No. 50466267 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges facilitate rugged, reproducible, and ultra-fast on-line solid phase extraction in compatible systems. Each features a water-wettable, mixed-mode polymeric sorbent optimized for high selectivity and sensitivity in extracting acidic compounds.
  • Designed for rugged, reproducible, and ultra-fast on-line solid phase extraction.
  • Contains 2.5 mg of water-wettable, mixed-mode polymeric sorbent.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Packaged with 96 cartridges per tray, directly loadable into automatic exchange or feeder units.
  • Offers both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity.
  • Enables sample concentration up to 15x without evaporation or reconstitution with an μElution plate.
Catalog No. 50-466-267 Supplier Waters Corp Supplier No. 186005783
These cartridges facilitate rugged, reproducible, and ultra-fast on-line solid phase extraction in compatible systems. Each features a water-wettable, mixed-mode polymeric sorbent optimized for high selectivity and sensitivity in extracting acidic compounds.
  • Designed for rugged, reproducible, and ultra-fast on-line solid phase extraction.
  • Contains 2.5 mg of water-wettable, mixed-mode polymeric sorbent.
  • Optimized for high selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Packaged with 96 cartridges per tray, directly loadable into automatic exchange or feeder units.
  • Offers both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity.
  • Enables sample concentration up to 15x without evaporation or reconstitution with an μElution plate.