Learn More
Waters Corp Waters Oasis MAX Online Prospekt-2/Symbiosis Cartridge, 80Å, 30 μm, 10 mm
Catalog No. 50466021 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Oasis MAX cartridges are designed for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE) in Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 10.4 mg of a water-wettable, mixed-mode polymeric sorbent optimized for extracting acidic compounds with anion-exchange groups, achieving higher selectivity and sensitivity.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE in Spark Holland Symbiosis and Prospekt-2 systems.
  • Each cartridge contains 10.4 mg of a water-wettable, mixed-mode polymeric sorbent, optimized for extracting acidic compounds.
  • Water-wettable sorbent maintains increased retention and superior recoveries even if it runs dry.
  • Flangeless design allows for dense packing in specialized automated SPE apparatus.
Catalog No. 50-466-021 Supplier Waters Corp Supplier No. 186004654
These Oasis MAX cartridges are designed for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE) in Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 10.4 mg of a water-wettable, mixed-mode polymeric sorbent optimized for extracting acidic compounds with anion-exchange groups, achieving higher selectivity and sensitivity.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE in Spark Holland Symbiosis and Prospekt-2 systems.
  • Each cartridge contains 10.4 mg of a water-wettable, mixed-mode polymeric sorbent, optimized for extracting acidic compounds.
  • Water-wettable sorbent maintains increased retention and superior recoveries even if it runs dry.
  • Flangeless design allows for dense packing in specialized automated SPE apparatus.