Learn More
Waters Corp Oasis MAX Online Column, 80Å, 30 μm, 2.1 mm x 20 mm
Supplier:  Waters Corp 186002052
Encompass_Preferred
The Oasis MAX online column is engineered for rugged, reproducible, and ultra-fast Solid Phase Extraction (SPE). It incorporates a water-wettable, mixed-mode polymeric sorbent for enhanced selectivity and sensitivity in extracting acidic compounds with anion-exchange groups. It is compatible with narrow-bore analytical columns.
  • Engineered for rugged, reproducible, ultra-fast online SPE.
  • Features water-wettable, mixed-mode polymeric sorbent for enhanced selectivity with acidic compounds.
  • Stable from pH 0 to 14.
  • Water-wettability ensures excellent retention even if sorbent bed dries.
  • Separates analytes from complex samples into acidic and basic/neutral fractions.
  • Compatible with narrow-bore analytical columns.
  • Suitable for online analysis systems and lab equipment.
  • Demonstrates excellent long-term reproducibility.
Catalog No. 50-780-908