Learn More
Waters Corp Oasis Online Column, 80Å, 30 μm, 3.9 mm X 20 mm
Catalog No. 50780910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX on-line column is engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE). It incorporates the Oasis MAX sorbent, a water-wettable, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting acidic compounds in anion-exchange groups.
  • Engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE).
  • Contains a water-wettable, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Designed for use with appropriate narrow-bore analytical columns.
  • Cartridge column features traditional HPLC column fittings and hardware.
Catalog No. 50-780-910 Supplier Waters Corp Supplier No. 186002054
The Oasis MAX on-line column is engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE). It incorporates the Oasis MAX sorbent, a water-wettable, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting acidic compounds in anion-exchange groups.
  • Engineered for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE).
  • Contains a water-wettable, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Designed for use with appropriate narrow-bore analytical columns.
  • Cartridge column features traditional HPLC column fittings and hardware.