Learn More
Waters Corp Waters Online Column, Oasis MAX, 30 μm, 3 mm, 20 mm
Supplier:  Waters Corp 186002053
Encompass_Preferred
The Oasis MAX on-line column is engineered for rugged, reproducible, and ultra-fast Solid Phase Extraction (SPE). It utilizes a water-wettable, mixed-mode polymeric sorbent for high selectivity in extracting acidic compounds. This column allows efficient separation of complex mixtures into acidic and basic/neutral fractions, ensuring consistent performance even if the sorbent bed dries.
  • Rugged, reproducible, ultra-fast on-line SPE.
  • Water-wettable, mixed-mode polymeric sorbent for selective extraction of acidic compounds.
  • Efficiently separates complex mixtures into acidic and basic/neutral fractions.
  • Maintains retention capacity, even if the sorbent bed runs dry.
  • Manufactured in a cGMP, ISO 9001 certified facility with rigorous batch testing.
  • Includes Performance Chromatogram and Certificate of Batch Analysis.
Catalog No. 50-780-909