Learn More
Waters Corp Waters Corporation 96-well Plate, Oasis MAX Mixed-Mode, 10 mg Sorbent, 30 μm
Catalog No. 50780199 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX 96-well plate features a novel, mixed-mode polymeric sorbent optimized for enhanced selectivity and sensitivity in extracting acidic compounds using anion-exchange groups. Its unique balance of hydrophobicity and water-wettability ensures consistent results even if individual wells dry out before sample loading. This plate is compatible with various manifold configurations and most robotic liquid handling systems.
  • Novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Unique balance of hydrophobicity and water-wettability, preventing poor results if wells dry out prior to sample loading.
  • Designed for use with many manifold configurations and most robotic liquid handling systems.
Catalog No. 50-780-199 Supplier Waters Corp Supplier No. 186000375
The Oasis MAX 96-well plate features a novel, mixed-mode polymeric sorbent optimized for enhanced selectivity and sensitivity in extracting acidic compounds using anion-exchange groups. Its unique balance of hydrophobicity and water-wettability ensures consistent results even if individual wells dry out before sample loading. This plate is compatible with various manifold configurations and most robotic liquid handling systems.
  • Novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Unique balance of hydrophobicity and water-wettability, preventing poor results if wells dry out prior to sample loading.
  • Designed for use with many manifold configurations and most robotic liquid handling systems.