Learn More
Waters Corp Waters 96-well Plate, Mixed-mode Sorbent, 30 mg/well, 30 μm
Catalog No. 50780198 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX 96-well plate is designed for solid phase extraction (SPE), featuring a novel, mixed-mode polymeric sorbent. It offers high selectivity and sensitivity for extracting acidic compounds. The plate is compatible with various manifold configurations and robotic liquid handling systems for automated, high-throughput SPE.
  • Unique hydrophobicity and water-wettability prevent poor results if wells dry out.
  • Stable from pH 0 to 14.
  • Enables separation of analytes from complex samples into acidic and basic/neutral compounds.
  • Dual modes of retention (reversed-phase and ion exchange) provide greater cleanup selectivity and sensitivity.
  • Offers 0.25 meq/g of quaternary-amine-ion-exchange capacity.
  • Innovative two-stage design for reliable high throughput SPE.
Catalog No. 50-780-198 Supplier Waters Corp Supplier No. 186000373
The Oasis MAX 96-well plate is designed for solid phase extraction (SPE), featuring a novel, mixed-mode polymeric sorbent. It offers high selectivity and sensitivity for extracting acidic compounds. The plate is compatible with various manifold configurations and robotic liquid handling systems for automated, high-throughput SPE.
  • Unique hydrophobicity and water-wettability prevent poor results if wells dry out.
  • Stable from pH 0 to 14.
  • Enables separation of analytes from complex samples into acidic and basic/neutral compounds.
  • Dual modes of retention (reversed-phase and ion exchange) provide greater cleanup selectivity and sensitivity.
  • Offers 0.25 meq/g of quaternary-amine-ion-exchange capacity.
  • Innovative two-stage design for reliable high throughput SPE.