Learn More
Waters Corp Oasis MAX 96-well Plate, 60 mg Sorbent per Well, 30 µm, 1/pk
Catalog No. 50780630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX 96-well plate contains the Oasis MAX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting acidic compounds with anion-exchange groups. The unique balance of hydrophobicity and water-wettability of the Oasis MAX sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis MAX 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.
Catalog No. 50-780-630 Supplier Waters Corp Supplier No. 186001256
The Oasis MAX 96-well plate contains the Oasis MAX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting acidic compounds with anion-exchange groups. The unique balance of hydrophobicity and water-wettability of the Oasis MAX sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis MAX 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.