Learn More
Waters Corp Waters Oasis MAX 96-well Plate, 60 mg Sorbent per Well, 60 μm
Catalog No. 50780629 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX 96-well plate utilizes a mixed-mode polymeric sorbent designed for the extraction of acidic compounds with anion-exchange groups, offering enhanced selectivity and sensitivity. Its balanced hydrophobic and water-wettable properties prevent issues if wells dry out before sample loading. A 60 μm particle size is recommended for viscous samples, and the plate is compatible with various manifold configurations and robotic liquid handling systems.
  • Optimized for selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Balanced hydrophobicity and water-wettability prevents issues if wells dry out before sample loading.
  • Compatible with multiple manifold configurations and most robotic liquid handling systems.
  • Supports high resolution, sensitivity, and throughput.
  • Reduces time and cost per sample while maintaining high-quality results.
Catalog No. 50-780-629 Supplier Waters Corp Supplier No. 186001205
The Oasis MAX 96-well plate utilizes a mixed-mode polymeric sorbent designed for the extraction of acidic compounds with anion-exchange groups, offering enhanced selectivity and sensitivity. Its balanced hydrophobic and water-wettable properties prevent issues if wells dry out before sample loading. A 60 μm particle size is recommended for viscous samples, and the plate is compatible with various manifold configurations and robotic liquid handling systems.
  • Optimized for selectivity and sensitivity in extracting acidic compounds with anion-exchange groups.
  • Balanced hydrophobicity and water-wettability prevents issues if wells dry out before sample loading.
  • Compatible with multiple manifold configurations and most robotic liquid handling systems.
  • Supports high resolution, sensitivity, and throughput.
  • Reduces time and cost per sample while maintaining high-quality results.