Learn More
Waters Corp Waters Oasis Plus Short Cartridge, 225 mg Sorbent, 60 μm, Mixed-Mode Polymeric
Catalog No. 50781817 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Plus Short Cartridge is designed for streamlined sample preparation, utilizing a novel, water-wettable, mixed-mode polymeric sorbent. It ensures high selectivity and sensitivity for acidic compounds and maintains retention even if dry. The 60 μm particle size is ideal for viscous samples. It supports various flow methods, extracts analytes from complex samples, and removes phospholipids to reduce ion suppression.
  • Utilizes a novel, mixed-mode polymeric sorbent with anion-exchange groups.
  • Maintains retention and excellent recoveries, even if the sorbent runs dry.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds.
  • Supports positive-pressure flow, vacuum-assisted devices, and automated systems.
  • Removes phospholipids and lysophospholipids, reducing ion suppression in LC/MS/MS analysis.
Catalog No. 50-781-817 Supplier Waters Corp Supplier No. 186003517
This Plus Short Cartridge is designed for streamlined sample preparation, utilizing a novel, water-wettable, mixed-mode polymeric sorbent. It ensures high selectivity and sensitivity for acidic compounds and maintains retention even if dry. The 60 μm particle size is ideal for viscous samples. It supports various flow methods, extracts analytes from complex samples, and removes phospholipids to reduce ion suppression.
  • Utilizes a novel, mixed-mode polymeric sorbent with anion-exchange groups.
  • Maintains retention and excellent recoveries, even if the sorbent runs dry.
  • Optimized for higher selectivity and sensitivity in extracting acidic compounds.
  • Supports positive-pressure flow, vacuum-assisted devices, and automated systems.
  • Removes phospholipids and lysophospholipids, reducing ion suppression in LC/MS/MS analysis.