Learn More
Waters Corp WATERS CORPORATION Oasis MAX Vac RC Cartridge, 30 mg Sorbent, 30 μm, 20 cc
Catalog No. 50780197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MAX Vac RC Cartridge features a Mixed-Mode Anion eXchange sorbent designed for acids. It is built upon the unique water-wettable Oasis HLB Copolymer, providing both reversed-phase and ion exchange modes of retention for enhanced cleanup selectivity and sensitivity for acidic compounds.
  • Fastest and simplest methods leading to the cleanest samples.
  • Highest sensitivity for small molecules and therapeutic peptides.
  • Reduced matrix effects for LC-MS/MS analyses.
  • Unsurpassed selectivity.
  • Highest solid-phase extraction recovery.
  • Exceptional product quality and reproducibility.
  • Wide selection of sorbents and a broad array of formats.
  • Ultra-clean syringe barrel and frits.
Catalog No. 50-780-197 Supplier Waters Corp Supplier No. 186000372
The Oasis MAX Vac RC Cartridge features a Mixed-Mode Anion eXchange sorbent designed for acids. It is built upon the unique water-wettable Oasis HLB Copolymer, providing both reversed-phase and ion exchange modes of retention for enhanced cleanup selectivity and sensitivity for acidic compounds.
  • Fastest and simplest methods leading to the cleanest samples.
  • Highest sensitivity for small molecules and therapeutic peptides.
  • Reduced matrix effects for LC-MS/MS analyses.
  • Unsurpassed selectivity.
  • Highest solid-phase extraction recovery.
  • Exceptional product quality and reproducibility.
  • Wide selection of sorbents and a broad array of formats.
  • Ultra-clean syringe barrel and frits.