Learn More
Waters Corp Waters Vac RC Cartridge, Oasis MAX, 20 cc, 60 mg, 30 μm
Catalog No. 50780196 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Vac RC cartridges feature Oasis MAX sorbent, a mixed-mode polymeric sorbent for extracting acidic compounds. Its water wettability ensures high retention and recovery even when dry. An integral 20 cc funnel-shaped reservoir aids processing larger volumes, and the cartridges are compatible with robotic sample preparation equipment and vacuum manifolds.
  • Mixed-mode polymeric sorbent for acidic compound extraction
  • Water wettable sorbent for consistent performance
  • 20 cc funnel-shaped reservoir
  • Compatible with robotic systems and vacuum manifolds
  • Ion exchange capacity: 0.2 meq/gram
  • Mass spectrometry compatible
  • Particle size: 30 μm
  • Pore size: 80 Å
  • Sorbent weight: 60 mg
  • pH range: 0 to 14
  • Material: Polypropylene
Catalog No. 50-780-196 Supplier Waters Corp Supplier No. 186000371
These Vac RC cartridges feature Oasis MAX sorbent, a mixed-mode polymeric sorbent for extracting acidic compounds. Its water wettability ensures high retention and recovery even when dry. An integral 20 cc funnel-shaped reservoir aids processing larger volumes, and the cartridges are compatible with robotic sample preparation equipment and vacuum manifolds.
  • Mixed-mode polymeric sorbent for acidic compound extraction
  • Water wettable sorbent for consistent performance
  • 20 cc funnel-shaped reservoir
  • Compatible with robotic systems and vacuum manifolds
  • Ion exchange capacity: 0.2 meq/gram
  • Mass spectrometry compatible
  • Particle size: 30 μm
  • Pore size: 80 Å
  • Sorbent weight: 60 mg
  • pH range: 0 to 14
  • Material: Polypropylene