Learn More
Waters Corp Waters Corporation μElution Plate, 96-well, MCX, 2 mg, 30 μm
Catalog No. 50818608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well μElution plate is engineered for Solid Phase Extraction (SPE) cleanup and analyte enrichment of sample volumes from 10 μL to 375 μL. It features a novel, mixed-mode polymeric sorbent (MCX) for high selectivity and sensitivity, allowing for 15-fold sample enrichment with 25 μL elution, and supports automated, high-throughput SPE.
  • Designed for SPE cleanup and analyte enrichment of sample volumes ranging from 10 μL to 375 μL.
  • Optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups.
  • Enabled elution in only 25 μL, providing sensitive, robust, and reproducible results without evaporation and reconstitution.
  • Compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Provides dual modes of retention: ion exchange and reversed-phase.
  • Stable from pH 0 to 14.
Catalog No. 50-818-608 Supplier Waters Corp Supplier No. 186001830BA
This 96-well μElution plate is engineered for Solid Phase Extraction (SPE) cleanup and analyte enrichment of sample volumes from 10 μL to 375 μL. It features a novel, mixed-mode polymeric sorbent (MCX) for high selectivity and sensitivity, allowing for 15-fold sample enrichment with 25 μL elution, and supports automated, high-throughput SPE.
  • Designed for SPE cleanup and analyte enrichment of sample volumes ranging from 10 μL to 375 μL.
  • Optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups.
  • Enabled elution in only 25 μL, providing sensitive, robust, and reproducible results without evaporation and reconstitution.
  • Compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Provides dual modes of retention: ion exchange and reversed-phase.
  • Stable from pH 0 to 14.