Learn More
Waters Corp Oasis MCX Bulk Media, 250 g, 30 μm, Mixed-Mode
Catalog No. 502391427 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product contains a 250 g bulk Oasis MCX SPE sorbent, 30 μm. Oasis MCX is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. Since the Oasis MCX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading.
  • Novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
  • Water wettable, maintains capability for higher retention and excellent recoveries even if the sorbent runs dry.
Catalog No. 50-239-1427 Supplier Waters Corp Supplier No. 186007552
Add to Cart
Add to Cart
Request a Quote
This product contains a 250 g bulk Oasis MCX SPE sorbent, 30 μm. Oasis MCX is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. Since the Oasis MCX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading.
  • Novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
  • Water wettable, maintains capability for higher retention and excellent recoveries even if the sorbent runs dry.