Learn More
Waters Corp Waters Oasis MCX Vac Cartridge, Mixed-Mode Polymeric, 1 cc, 30 mg, 30 μm
Catalog No. 50819042 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc Oasis MCX cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. Water-wettable for consistent performance, they simplify sample preparation and are ideal for vacuum manifolds and automated SPE instruments.
  • High selectivity and sensitivity for basic compounds.
  • Water-wettable design ensures consistent performance even if dry.
  • Dual modes of retention: ion exchange and reversed-phase.
  • Broad pH stability (0-14) allows flexible method development.
  • Superior binding capacity vs. other SPE sorbents.
  • Prevents analyte breakthrough or recovery loss.
  • 1.0 meq/g sulfonic acid ion exchange capacity.
Catalog No. 50-819-042 Supplier Waters Corp Supplier No. 186000252
These 1 cc Oasis MCX cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. Water-wettable for consistent performance, they simplify sample preparation and are ideal for vacuum manifolds and automated SPE instruments.
  • High selectivity and sensitivity for basic compounds.
  • Water-wettable design ensures consistent performance even if dry.
  • Dual modes of retention: ion exchange and reversed-phase.
  • Broad pH stability (0-14) allows flexible method development.
  • Superior binding capacity vs. other SPE sorbents.
  • Prevents analyte breakthrough or recovery loss.
  • 1.0 meq/g sulfonic acid ion exchange capacity.