Learn More
Waters Corp Waters Corporation Solid Phase Extraction Vac Cartridge, Oasis MCX, 3 cc, 60 mg, 60 μm
Catalog No. 50818662 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MCX 3 cc Vac Cartridge features a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in basic compound extraction. It is water wettable, ensuring consistent recoveries even if it runs dry. Designed for viscous samples (60 μm particle size), it is compatible with vacuum manifolds and automated SPE. Offers pH stability from 0 to 14 and higher binding capacity than silica-based sorbents, facilitating method development.
  • Optimized for high selectivity & sensitivity in basic compound extraction, water wettable for consistent retention & recovery when dry.
  • 60 μm particle size suitable for viscous samples; compatible with vacuum manifolds & automated SPE.
  • Novel mixed-mode polymeric sorbent provides pH stability from 0 to 14 & greater binding capacity than silica-based SPE, preventing breakthrough/recovery loss.
Catalog No. 50-818-662 Supplier Waters Corp Supplier No. 186000253
Oasis MCX 3 cc Vac Cartridge features a novel, mixed-mode polymeric sorbent for high selectivity and sensitivity in basic compound extraction. It is water wettable, ensuring consistent recoveries even if it runs dry. Designed for viscous samples (60 μm particle size), it is compatible with vacuum manifolds and automated SPE. Offers pH stability from 0 to 14 and higher binding capacity than silica-based sorbents, facilitating method development.
  • Optimized for high selectivity & sensitivity in basic compound extraction, water wettable for consistent retention & recovery when dry.
  • 60 μm particle size suitable for viscous samples; compatible with vacuum manifolds & automated SPE.
  • Novel mixed-mode polymeric sorbent provides pH stability from 0 to 14 & greater binding capacity than silica-based SPE, preventing breakthrough/recovery loss.