Learn More
Waters Corp Waters Oasis Vac Cartridge, 3 cc, 60 mg Sorbent, 30 μm
Catalog No. 50818663 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MCX 3 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for extracting basic compounds with cation-exchange groups. Water-wettable properties ensure high retention and excellent recoveries, even if the sorbent runs dry. These cartridges are compatible with vacuum manifolds and automated SPE instruments.
  • Novel, mixed-mode polymeric sorbent for basic compounds.
  • Water-wettable for consistent performance even if dry.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Offers dual retention modes: ion-exchange and reversed-phase.
  • Stable across pH 0 to 14 range.
  • Facilitates high SPE recovery and clean extracts.
Catalog No. 50-818-663 Supplier Waters Corp Supplier No. 186000254
Oasis MCX 3 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for extracting basic compounds with cation-exchange groups. Water-wettable properties ensure high retention and excellent recoveries, even if the sorbent runs dry. These cartridges are compatible with vacuum manifolds and automated SPE instruments.
  • Novel, mixed-mode polymeric sorbent for basic compounds.
  • Water-wettable for consistent performance even if dry.
  • Compatible with vacuum manifolds and automated SPE instruments.
  • Offers dual retention modes: ion-exchange and reversed-phase.
  • Stable across pH 0 to 14 range.
  • Facilitates high SPE recovery and clean extracts.