Learn More
Waters Corp Waters Oasis MCX Vac Cartridge, 1 cc, 30 mg Sorbent, 60 μm
Catalog No. 50780485 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. The water-wettable sorbent ensures high retention and excellent recoveries even if it runs dry, removing the need to prevent sorbent bed dry-out. Designed as syringe-barrel-type cartridges, they are suitable for vacuum manifolds and automated SPE instruments.
  • Mixed-mode polymeric sorbent, cation-exchange groups
  • High selectivity and sensitivity for basic compounds
  • Water-wettable, maintains retention and recovery even if dried
  • Eliminates need for sorbent dry-out prevention
  • 60 μm particle size for viscous samples
  • Dual retention modes: ion exchange and reversed-phase
  • Stable across wide operating pH range (0 to 14)
  • Maximizes SPE recovery; provides clean extracts
Catalog No. 50-780-485 Supplier Waters Corp Supplier No. 186000782
These 1 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. The water-wettable sorbent ensures high retention and excellent recoveries even if it runs dry, removing the need to prevent sorbent bed dry-out. Designed as syringe-barrel-type cartridges, they are suitable for vacuum manifolds and automated SPE instruments.
  • Mixed-mode polymeric sorbent, cation-exchange groups
  • High selectivity and sensitivity for basic compounds
  • Water-wettable, maintains retention and recovery even if dried
  • Eliminates need for sorbent dry-out prevention
  • 60 μm particle size for viscous samples
  • Dual retention modes: ion exchange and reversed-phase
  • Stable across wide operating pH range (0 to 14)
  • Maximizes SPE recovery; provides clean extracts