Learn More
Waters Corp Waters Oasis MCX Vac Cartridge, 1 cc, 10 mg, 30 μm
Supplier:  Waters Corp 186004648
Encompass_Preferred
The Oasis MCX 1 cc Vac Cartridge features a novel, mixed-mode polymeric sorbent, optimized for enhanced selectivity and sensitivity in extracting basic compounds. Its water-wettable nature ensures consistent retention and excellent recoveries, even if the sorbent runs dry, simplifying sample preparation. Cartridges are designed for use with vacuum manifolds and automated solid-phase extraction (SPE) instruments.
  • Novel, mixed-mode polymeric sorbent optimized for high selectivity and sensitivity in extracting basic compounds with cation-exchange groups.
  • Water wettable, maintaining retention and excellent recoveries even if sorbent runs dry.
  • Designed for use with vacuum manifolds and automated SPE instruments.
  • Enables separation of analytes from complex samples into acidic and basic or neutral compounds.
  • Known for greater recovery, repeatability, stronger retention, and selectivity.
Catalog No. 50-791-170