Learn More
Waters Corp Oasis MCX Cartridge, 1 cc/30 mg, Gilson Adapter, 30 μm
Catalog No. 50780789 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MCX (Mixed-Mode Cation eXchange) Sorbent is a water-wettable polymeric sorbent stable from pH 0–14. It is designed for selective retention of basic drugs using cation-exchange groups. This mixed-mode solid-phase extraction sorbent provides dual retention, leading to high specificity, increased sensitivity, and cleaner extracts by reducing matrix effects. Effective for enriching analytes, it's recommended for weak bases with a pKa between 2 and 10.
  • Tightly controlled ion-exchange capacity of 1 meq/g.
  • No silanol groups complicate retention or method development.
  • Provides orthogonality and selectivity.
  • Offers two screening protocols.
  • Captures and concentrates basic compounds while removing matrix interferences.
  • Uses 3-step or 4-step methods; 3-step for cleaner results, 4-step for additional matrix removal.
Catalog No. 50-780-789 Supplier Waters Corp Supplier No. 186001888
The Oasis MCX (Mixed-Mode Cation eXchange) Sorbent is a water-wettable polymeric sorbent stable from pH 0–14. It is designed for selective retention of basic drugs using cation-exchange groups. This mixed-mode solid-phase extraction sorbent provides dual retention, leading to high specificity, increased sensitivity, and cleaner extracts by reducing matrix effects. Effective for enriching analytes, it's recommended for weak bases with a pKa between 2 and 10.
  • Tightly controlled ion-exchange capacity of 1 meq/g.
  • No silanol groups complicate retention or method development.
  • Provides orthogonality and selectivity.
  • Offers two screening protocols.
  • Captures and concentrates basic compounds while removing matrix interferences.
  • Uses 3-step or 4-step methods; 3-step for cleaner results, 4-step for additional matrix removal.