Learn More
Waters Corp Oasis Vac Cartridge, MCX, 6 cc, 150 mg, 60 μm
Catalog No. 50780151 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 6 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds with cation-exchange groups. Its water wettable nature ensures excellent recoveries even when run dry. With a 60 μm particle size, it is ideal for viscous samples and is compatible with vacuum manifolds and automated SPE instruments.
  • Optimized for high selectivity and sensitivity in basic compound extraction.
  • Water wettable sorbent maintains excellent recoveries even when run dry.
  • Strong mixed-mode cation exchange polymeric sorbent surpasses traditional silica-based sorbents.
  • Offers dual modes of retention (ion exchange, reversed-phase) across a 0 to 14 pH range.
  • Provides superior binding capacity: 1.0 meq/g sulfonic acid ion exchange capacity.
  • Simplifies method development and enhances analytical system performance.
Catalog No. 50-780-151 Supplier Waters Corp Supplier No. 186000255
These 6 cc cartridges feature a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds with cation-exchange groups. Its water wettable nature ensures excellent recoveries even when run dry. With a 60 μm particle size, it is ideal for viscous samples and is compatible with vacuum manifolds and automated SPE instruments.
  • Optimized for high selectivity and sensitivity in basic compound extraction.
  • Water wettable sorbent maintains excellent recoveries even when run dry.
  • Strong mixed-mode cation exchange polymeric sorbent surpasses traditional silica-based sorbents.
  • Offers dual modes of retention (ion exchange, reversed-phase) across a 0 to 14 pH range.
  • Provides superior binding capacity: 1.0 meq/g sulfonic acid ion exchange capacity.
  • Simplifies method development and enhances analytical system performance.