Learn More
Waters Corp Waters Oasis MCX Vac Cartridge, 6 cc, 500 mg, 60 μm
Catalog No. 50780482 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 6 cc cartridges utilize a novel, mixed-mode polymeric sorbent for optimal extraction of basic compounds via cation-exchange groups. The water-wettable sorbent maintains high retention and recoveries even if run dry, eliminating the need for special drying prevention steps. With a 60 μm particle size, they are ideal for viscous samples. Designed for vacuum manifolds and automated SPE instruments, these cartridges offer dual retention modes (ion exchange and reversed-phase) on a stable organic co-polymer from pH 0–14.
  • Optimized for high selectivity in basic compound extraction.
  • Water-wettable sorbent ensures high retention and recovery dry.
  • Dual retention modes: ion exchange and reversed-phase.
  • Clean, stable, high-surface-area organic co-polymer sorbent.
  • 60 μm particle size for viscous samples.
  • Syringe-barrel design for vacuum and automated SPE.
Catalog No. 50-780-482 Supplier Waters Corp Supplier No. 186000776
These 6 cc cartridges utilize a novel, mixed-mode polymeric sorbent for optimal extraction of basic compounds via cation-exchange groups. The water-wettable sorbent maintains high retention and recoveries even if run dry, eliminating the need for special drying prevention steps. With a 60 μm particle size, they are ideal for viscous samples. Designed for vacuum manifolds and automated SPE instruments, these cartridges offer dual retention modes (ion exchange and reversed-phase) on a stable organic co-polymer from pH 0–14.
  • Optimized for high selectivity in basic compound extraction.
  • Water-wettable sorbent ensures high retention and recovery dry.
  • Dual retention modes: ion exchange and reversed-phase.
  • Clean, stable, high-surface-area organic co-polymer sorbent.
  • 60 μm particle size for viscous samples.
  • Syringe-barrel design for vacuum and automated SPE.