Learn More
Waters Corp Waters Vac Cartridge, 20 cc, 1 g, 60 μm MCX
Catalog No. 50818664 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 20 cc Oasis MCX cartridges contain a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. This water-wettable sorbent ensures high retention and excellent recoveries even if dry, eliminating extra precautions. The 60 μm particle size suits viscous samples. These cartridges are for vacuum manifolds and automated SPE instruments.
  • Novel, mixed-mode polymeric sorbent for enhanced selectivity and sensitivity
  • Optimized for extracting basic compounds with cation-exchange groups
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry
  • Eliminates the need for extra precautions to prevent sorbent beds from drying out
  • 60 μm particle size is ideal for viscous samples
  • Designed for use with vacuum manifolds and automated SPE instruments
Catalog No. 50-818-664 Supplier Waters Corp Supplier No. 186000777
The 20 cc Oasis MCX cartridges contain a novel, mixed-mode polymeric sorbent, optimized for high selectivity and sensitivity in extracting basic compounds. This water-wettable sorbent ensures high retention and excellent recoveries even if dry, eliminating extra precautions. The 60 μm particle size suits viscous samples. These cartridges are for vacuum manifolds and automated SPE instruments.
  • Novel, mixed-mode polymeric sorbent for enhanced selectivity and sensitivity
  • Optimized for extracting basic compounds with cation-exchange groups
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry
  • Eliminates the need for extra precautions to prevent sorbent beds from drying out
  • 60 μm particle size is ideal for viscous samples
  • Designed for use with vacuum manifolds and automated SPE instruments