Learn More
Waters Corp Waters SPE Cartridge, MCX, 1 cc, Flangeless, 30 mg Sorbent
Catalog No. 50780785 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flangeless vac cartridge features a novel, mixed-mode polymeric sorbent optimized for extracting basic compounds with cation-exchange groups. It provides high selectivity, sensitivity, and excellent recoveries even if the sorbent runs dry due to its water-wettable nature. Designed for automated SPE systems, it offers a broad operating pH range of 0 to 14 pH.
  • Contains a novel, mixed-mode polymeric sorbent
  • Optimized for extracting basic compounds with cation-exchange groups
  • Maintains high retention and excellent recoveries even if sorbent runs dry
  • Flangeless configuration for close packing in automated systems
  • Stable across a broad pH range, enabling robust processes
  • Utilizes ion exchange and reversed-phase retention modes
  • Operates within a pH range of 0 to 14 pH
  • Provides high SPE recovery, clean extracts, and low method variability
Catalog No. 50-780-785 Supplier Waters Corp Supplier No. 186001881
This flangeless vac cartridge features a novel, mixed-mode polymeric sorbent optimized for extracting basic compounds with cation-exchange groups. It provides high selectivity, sensitivity, and excellent recoveries even if the sorbent runs dry due to its water-wettable nature. Designed for automated SPE systems, it offers a broad operating pH range of 0 to 14 pH.
  • Contains a novel, mixed-mode polymeric sorbent
  • Optimized for extracting basic compounds with cation-exchange groups
  • Maintains high retention and excellent recoveries even if sorbent runs dry
  • Flangeless configuration for close packing in automated systems
  • Stable across a broad pH range, enabling robust processes
  • Utilizes ion exchange and reversed-phase retention modes
  • Operates within a pH range of 0 to 14 pH
  • Provides high SPE recovery, clean extracts, and low method variability