Learn More
Waters Corp Waters Flangeless Vac Cartridge, Oasis MCX, 3 cc, 60 mg, 30 μm
Catalog No. 50780786 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MCX Flangeless Vac Cartridge is designed for solid phase extraction of basic compounds. It features a novel, mixed-mode polymeric sorbent that is water wettable, ensuring consistent performance even if the sorbent runs dry. Its flangeless design is ideal for close packing and processing in specialized automated SPE systems.
  • Novel, mixed-mode polymeric sorbent for higher selectivity and sensitivity.
  • Extracts basic compounds using cation-exchange groups.
  • Water-wettable sorbent ensures high retention and recoveries.
  • Flangeless design allows close packing in automated SPE systems.
  • 3 cc barrel size with 60 mg sorbent weight.
  • 30 μm particle size.
  • Effective across a pH range of 0 to 14.
  • Durable polypropylene construction.
  • Mass spectrometry compatible.
  • Ion exchange capacity of 1 meq/gram.
Catalog No. 50-780-786 Supplier Waters Corp Supplier No. 186001882
The Oasis MCX Flangeless Vac Cartridge is designed for solid phase extraction of basic compounds. It features a novel, mixed-mode polymeric sorbent that is water wettable, ensuring consistent performance even if the sorbent runs dry. Its flangeless design is ideal for close packing and processing in specialized automated SPE systems.
  • Novel, mixed-mode polymeric sorbent for higher selectivity and sensitivity.
  • Extracts basic compounds using cation-exchange groups.
  • Water-wettable sorbent ensures high retention and recoveries.
  • Flangeless design allows close packing in automated SPE systems.
  • 3 cc barrel size with 60 mg sorbent weight.
  • 30 μm particle size.
  • Effective across a pH range of 0 to 14.
  • Durable polypropylene construction.
  • Mass spectrometry compatible.
  • Ion exchange capacity of 1 meq/gram.