Learn More
Waters Corp Oasis Online Prospekt-2/Symbiosis Cartridge, MCX, 30 μm, 80 Å, 10 mm
Catalog No. 50466020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE) in the Spark Holland Symbiosis and Prospekt-2 systems. They contain 10.4 mg of a novel, mixed-mode polymeric sorbent, optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
  • Contains 10.4 mg of a novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups.
  • Each 800-series cartridge tray contains 96 cartridges.
  • Can be directly loaded into the Automatic Cartridge Exchange (ACE) or the ACE Feeder units of the Symbiosis/Prospekt-2 systems without the stainless steel tray holder/adaptor.
Catalog No. 50-466-020 Supplier Waters Corp Supplier No. 186004653
The cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line Solid Phase Extraction (SPE) in the Spark Holland Symbiosis and Prospekt-2 systems. They contain 10.4 mg of a novel, mixed-mode polymeric sorbent, optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
  • Contains 10.4 mg of a novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups.
  • Each 800-series cartridge tray contains 96 cartridges.
  • Can be directly loaded into the Automatic Cartridge Exchange (ACE) or the ACE Feeder units of the Symbiosis/Prospekt-2 systems without the stainless steel tray holder/adaptor.