Learn More
Waters Corp Oasis MCX On-line Cartridge, 30 μm, 1 x 10 mm
Catalog No. 50466266 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This mixed-mode cation exchange solid-phase extraction (SPE) cartridge is designed for the efficient extraction of basic compounds. Built upon a water-wettable copolymer, it offers both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity and sensitivity.
  • Mixed-mode cation exchange sorbent for bases (pKa 2–10).
  • Provides both reversed-phase and ion exchange modes of retention.
  • Built upon water-wettable HLB copolymer.
  • Offers greater cleanup selectivity and sensitivity for basic compounds.
  • Designed for on-line applications.
  • 30 μm particle size.
  • 1 x 10 mm dimensions.
Catalog No. 50-466-266 Supplier Waters Corp Supplier No. 186005782
This mixed-mode cation exchange solid-phase extraction (SPE) cartridge is designed for the efficient extraction of basic compounds. Built upon a water-wettable copolymer, it offers both reversed-phase and ion exchange retention modes for enhanced cleanup selectivity and sensitivity.
  • Mixed-mode cation exchange sorbent for bases (pKa 2–10).
  • Provides both reversed-phase and ion exchange modes of retention.
  • Built upon water-wettable HLB copolymer.
  • Offers greater cleanup selectivity and sensitivity for basic compounds.
  • Designed for on-line applications.
  • 30 μm particle size.
  • 1 x 10 mm dimensions.