Learn More
Waters Corp Waters Online Column, Oasis MCX, 80Å, 30 μm, 2.1 mm X 20 mm
Supplier:  Waters Corp 186002046
Encompass_Preferred
The Oasis MCX on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. It contains the Oasis MCX sorbent, a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups. This column is intended for use with appropriate narrow-bore analytical columns, and features traditional HPLC column fittings and hardware.
  • Contains Oasis MCX sorbent, a novel, mixed-mode polymeric sorbent.
  • Optimized for higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE.
  • Features traditional HPLC column fittings and hardware.
  • Designed for use with appropriate narrow-bore analytical columns.
Catalog No. 50-780-902